Learn More
Pickering Laboratories Inc Artificial Saliva, Modified Fusayama/Mayer, Custom pH up to 6.5, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. NC1873810
Encompass_Preferred
This variation of classic Fusayama/Meyer formulation contains Potassium Thiocyanate that is present in natural saliva and has been shown to be elevated in smokers. This formulation is manufactured according to customer’s pH specifications in the range of pH 3.0-6.5. The formulation contains a preservative to prevent bacteria growth and can be stored at room temperature
Catalog No. NC1873810 Supplier Pickering Laboratories Inc Supplier No. 17000315
This variation of classic Fusayama/Meyer formulation contains Potassium Thiocyanate that is present in natural saliva and has been shown to be elevated in smokers. This formulation is manufactured according to customer’s pH specifications in the range of pH 3.0-6.5. The formulation contains a preservative to prevent bacteria growth and can be stored at room temperature