Learn More
Pickering Laboratories Inc Artificial Sebum, ASTM D4265-14, Not Stabilized, each (25 g/bottle)
SDP
Supplier:  Pickering Laboratories Inc 17000700
Encompass_Preferred
Temperature Sensitive, Refrigerate. Sebum is an oily secretion produced by sebaceous glands, which spreads over the hair and skin for waterproofing purposes. Pickering Labs manufactures an artificial sebum formulation according to ASTM D4265-14 or ASTM D4265-98. It is ready-to-use and provides the reliability, reproducibility and convenience needed for testing. This formulation can be used according to AATCC Standard Test Method 130-2010 for evaluating the efficacy of home laundry products and conditions to remove stains from fabric
Catalog No. 50-135-4826