Learn More
Pickering Laboratories Inc Artificial Urine, DIN EN 1616:1999, Not Stabilized, each (19.8 L/carboy)
SDP
Catalog No. 501974233
Encompass_Preferred
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This formulation is prepared according to DIN EN 1616:1999 standard procedure. DIN EN 1616 specifies the method to test sterile urethral catheters. This ready-to-use solution should be stored frozen to avoid bacteria growth. The pH of the solution is 6.6
Catalog No. 50-197-4233 Supplier Pickering Laboratories Inc Supplier No. 17000603
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This formulation is prepared according to DIN EN 1616:1999 standard procedure. DIN EN 1616 specifies the method to test sterile urethral catheters. This ready-to-use solution should be stored frozen to avoid bacteria growth. The pH of the solution is 6.6