Learn More
Pickering Laboratories Inc Artificial Urine, pH 6.0, Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501354831
Encompass_Preferred
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This artificial urine is designed for testing metallic biomaterials used to produce urological implants and catheters. This convenient product is a ready-to-use solution. The formulation contains a non-toxic preservative to avoid bacteria growth and can be stored at room temperature. The pH of the final solution is 6.0
Catalog No. 50-135-4831 Supplier Pickering Laboratories Inc Supplier No. 17000016
Add to Cart
Add to Cart
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. This artificial urine is designed for testing metallic biomaterials used to produce urological implants and catheters. This convenient product is a ready-to-use solution. The formulation contains a non-toxic preservative to avoid bacteria growth and can be stored at room temperature. The pH of the final solution is 6.0