Learn More
Pickering Laboratories Inc Artificial Urine, Medium, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. NC1034753
Encompass_Preferred
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. The ready-to-use solution closely resembles composition of human urine and can be used for clinical studies as well as for product testing. This formulation supports growth of a wide range of urinary pathogens and it is also capable of forming crystals similar to these found in natural urinary tract infections. It can be used as negative controls in laboratory testing. pH of the Artificial Urine Medium is 6.5. This product is stored frozen
Catalog No. NC1034753 Supplier Pickering Laboratories Inc Supplier No. 17000018
Add to Cart
Add to Cart
Our Artificial Urine solutions are formulated both according to official testing methods as well as published research on the use of artificial urine for corrosion testing and related applications. The products do not come sterile but could be sterilized before use using filtration. The ready-to-use solution closely resembles composition of human urine and can be used for clinical studies as well as for product testing. This formulation supports growth of a wide range of urinary pathogens and it is also capable of forming crystals similar to these found in natural urinary tract infections. It can be used as negative controls in laboratory testing. pH of the Artificial Urine Medium is 6.5. This product is stored frozen