Learn More
Aviva Systems Biology Corp ARTN Antibody - middle region (ARP63920_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63920_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARTN. It was validated on Western Blot. . Alternate names: ENOVIN, EVN, NBNApplicationWB. Species reactivity: Human
Catalog No. 50-288-117