Learn More
Bioworld ARTOCARPUS INTEGRIFOLIA, 1MG
Supplier:  Bioworld 217610901
Encompass
Artocarpus integrifolia Lectin (AIA) - Cy3liquid, from jacalinSource: Artocarpus integrifolia (Jackfruit)Activity: Less than 0.5 ug /ml will agglutinate neuraminidase treated human erythrocytes.Protein Concentration (Based on OD 280): 1 mg purified AIA- Cy3 / 1 mL bufferCarbohydrate Specificity: Galß1-3GalNAcInhibitory Carbohydrate: Galactose or MelibioseDivalent ions required: NoneMitogenic Activity: YesConjugation: Cy3Excitation: Maximum at 550 nmEmission: Maximum at 570 nmFluorescence: Orange
Catalog No. NC2092184