Learn More
Bioworld ARTOCARPUS INTEGRIFOLIA LEC1MG
Catalog No. NC3315162
Encompass
Artocarpus integrifolia Lectin (AIA) is purified by affinity chromatography /SKU: 21510001/ and is tetrameric two-chain structure consisting of four identical protomers with an isoelectric point of pH 7.8. The lectin has specificity for blood group types O / pos.SA/ and T. Jacalin is considered a galactose-specific lectin and elutes with galactose or melibiose.
Catalog No. NC3315162 Supplier Bioworld Supplier No. 215100251
Artocarpus integrifolia Lectin (AIA) is purified by affinity chromatography /SKU: 21510001/ and is tetrameric two-chain structure consisting of four identical protomers with an isoelectric point of pH 7.8. The lectin has specificity for blood group types O / pos.SA/ and T. Jacalin is considered a galactose-specific lectin and elutes with galactose or melibiose.