Learn More
Sigma Aldrich Fine Chemicals Biosciences ARTR self sealing barrier
Catalog No. 502909093
Encompass
Precision-molded tips with the Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies forensics general PCR work and radioisotope sampling.Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase- DNase- DNA- and pyrogen-free.
Catalog No. 50-290-9093 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A25981CS
Precision-molded tips with the Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies forensics general PCR work and radioisotope sampling.Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase- DNase- DNA- and pyrogen-free.