Learn More
Sigma Aldrich Fine Chemicals Biosciences ARTR self-sealing barrier
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A27231PAK
Encompass
Precision-molded tips with the Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling.Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Catalog No. 50-289-1539