Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arundic acid | 185517-21-9 | MFCD08458423 | 98.0% | 186.29 | C11H22O2 | 25 MG
SDP

Catalog No. 5000240257
Encompass_Preferred

Arundic acid is an astrocyte-modulating small molecule used in neuroscience research to modulate astrocyte activation and related signaling pathways. It is supplied as a purified solid for research applications; users should follow the supplier datasheet for storage, handling, and formulation instructions for in vitro and in vivo use.

  • Purity typically 98.0%.
  • Molecular weight 186.29 g/mol.
  • Molecular formula C11H22O2.
  • Supplied as a dry solid suitable for dissolution in common solvents.
  • Suitable for both in vitro and in vivo experimental models.
  • Includes datasheet guidance for storage and handling.
  • Available in small mg-scale quantities for exploratory studies.

Catalog No. 50-002-40257 Supplier Medchemexpress LLC Supplier No. HY10766125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arundic acid is an astrocyte-modulating small molecule used in neuroscience research to modulate astrocyte activation and related signaling pathways. It is supplied as a purified solid for research applications; users should follow the supplier datasheet for storage, handling, and formulation instructions for in vitro and in vivo use.

  • Purity typically 98.0%.
  • Molecular weight 186.29 g/mol.
  • Molecular formula C11H22O2.
  • Supplied as a dry solid suitable for dissolution in common solvents.
  • Suitable for both in vitro and in vivo experimental models.
  • Includes datasheet guidance for storage and handling.
  • Available in small mg-scale quantities for exploratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.