Learn More
Medchemexpress LLC Arundic acid | 185517-21-9 | MFCD08458423 | 98.0% | 186.29 | C11H22O2 | 50 MG
SDP
Catalog No. 5000240287
Encompass_Preferred
Arundic acid (CAS 185517-21-9) is a small-molecule astrocyte-modulating research reagent (C11H22O2, MW 186.29) used in neuroscience studies to modulate astrocyte activation and signaling. It has been reported to delay expansion of cerebral infarcts and to inhibit synthesis of S-100β, and is provided as a high-purity compound for research and analytical applications.
  • Astrocyte-modulating activity that reduces astrocyte activation.
  • Evidence of delaying cerebral infarct expansion in preclinical studies.
  • Inhibits S-100β synthesis and influences glutamate transporter pathways.
  • Supplied as a high-purity research reagent (purity 98.0%).
  • Storage recommendations available for long-term stability.
Catalog No. 50-002-40287 Supplier Medchemexpress LLC Supplier No. HY10766150MG
Arundic acid (CAS 185517-21-9) is a small-molecule astrocyte-modulating research reagent (C11H22O2, MW 186.29) used in neuroscience studies to modulate astrocyte activation and signaling. It has been reported to delay expansion of cerebral infarcts and to inhibit synthesis of S-100β, and is provided as a high-purity compound for research and analytical applications.
  • Astrocyte-modulating activity that reduces astrocyte activation.
  • Evidence of delaying cerebral infarct expansion in preclinical studies.
  • Inhibits S-100β synthesis and influences glutamate transporter pathways.
  • Supplied as a high-purity research reagent (purity 98.0%).
  • Storage recommendations available for long-term stability.