Learn More
Medchemexpress LLC Arundic acid | 185517-21-9 | MFCD08458423 | 98.0% | 186.29 g/mol | C11H22O2 | 1 ML

Supplier: Medchemexpress LLC HY10766110MM 1ML
Arundic acid is a small-molecule astrocyte-modulating agent used in neuroscience research to investigate neurodegeneration and neurological disease mechanisms. It is available as a high-purity research chemical and as a ready-to-use solution for in vitro studies.
- Modulates astrocyte activity in preclinical research.
- Available as a solid and as a 10 mM solution in DMSO.
- High purity suitable for research (98.0%).
- Molecular formula C11H22O2; molecular weight 186.29 g/mol.
- Storage and handling information provided for both solid and solution forms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.