Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arundic acid | 185517-21-9 | MFCD08458423 | 98.0% | 186.29 g·mol⁻¹ | C11H22O2 | 100 MG
SDP

Catalog No. 5000240244
Encompass_Preferred

Arundic acid is a small-molecule astrocyte-modulating agent used in neuroscience research to study astrocyte activation and neuroprotective mechanisms. It is supplied as a purified solid for laboratory assays and chemical analysis; handle and store according to the product safety data sheet. Not for human or clinical use.

  • Astrocyte-modulating small molecule for neuroscience research.
  • High reported purity suitable for biochemical and cellular assays.
  • Molecular formula C11H22O2 and molecular weight 186.29 g·mol⁻1.
  • Available in multiple pack sizes including 100 mg for laboratory studies.
  • Storage and handling conditions provided in the safety data sheet.

Catalog No. 50-002-40244 Supplier Medchemexpress LLC Supplier No. HY107661100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arundic acid is a small-molecule astrocyte-modulating agent used in neuroscience research to study astrocyte activation and neuroprotective mechanisms. It is supplied as a purified solid for laboratory assays and chemical analysis; handle and store according to the product safety data sheet. Not for human or clinical use.

  • Astrocyte-modulating small molecule for neuroscience research.
  • High reported purity suitable for biochemical and cellular assays.
  • Molecular formula C11H22O2 and molecular weight 186.29 g·mol⁻1.
  • Available in multiple pack sizes including 100 mg for laboratory studies.
  • Storage and handling conditions provided in the safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.