Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arv-771 | 1949837-12-0 | MFCD30738017 | >95.0% | 986.64 g/mol | C49H60ClN9O7S2 | 1 ML
SDP

Catalog No. 5000211893
Encompass_Preferred

ARV-771 is a PROTAC BET degrader used in cell biology and cancer research to induce targeted degradation of BET family proteins. The compound is supplied as powder or as a prepared solution, is highly soluble in DMSO, and is provided with manufacturer guidance for preparing millimolar stock solutions. Molecular formula C49H60ClN9O7S2; molecular weight 986.64 g/mol; CAS 1949837-12-0.

  • Proteolysis-targeting chimera mechanism for BET degradation
  • Available as powder or in solvent for experimental flexibility
  • High solubility in DMSO (≥50 mg/mL) enables concentrated stocks
  • Stock preparation guidance supplied for 1, 5, and 10 mM concentrations
  • Storage recommendations for powder and in-solution forms
  • Formulation examples provided for in vivo dosing vehicles

Catalog No. 50-002-11893 Supplier Medchemexpress LLC Supplier No. HY10097210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARV-771 is a PROTAC BET degrader used in cell biology and cancer research to induce targeted degradation of BET family proteins. The compound is supplied as powder or as a prepared solution, is highly soluble in DMSO, and is provided with manufacturer guidance for preparing millimolar stock solutions. Molecular formula C49H60ClN9O7S2; molecular weight 986.64 g/mol; CAS 1949837-12-0.

  • Proteolysis-targeting chimera mechanism for BET degradation
  • Available as powder or in solvent for experimental flexibility
  • High solubility in DMSO (≥50 mg/mL) enables concentrated stocks
  • Stock preparation guidance supplied for 1, 5, and 10 mM concentrations
  • Storage recommendations for powder and in-solution forms
  • Formulation examples provided for in vivo dosing vehicles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.