Learn More
Medchemexpress LLC HY-16954 50mg , ARV-825 CAS:1818885-28-7 Purity:98%
SDP
Catalog No. 501493618
Encompass_Preferred
Medchemexpress, HY-16954 50mg ARV-825 CAS:1818885-28-7 Formula:C46H47ClN8O9S Kd: 90 nM (Bromodomain 1 of BRD4), 28 nM (Bromodomain 2 of BRD4) Purity:98% ARV-825 is a BRD4 Inhibitor based on PROTAC technology. ARV-825 shows affinity to BD1 and BD2 of BRD4 with K d s of 90 and 28 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3618 Supplier Medchemexpress LLC Supplier No. HY1695450MG
Medchemexpress, HY-16954 50mg ARV-825 CAS:1818885-28-7 Formula:C46H47ClN8O9S Kd: 90 nM (Bromodomain 1 of BRD4), 28 nM (Bromodomain 2 of BRD4) Purity:98% ARV-825 is a BRD4 Inhibitor based on PROTAC technology. ARV-825 shows affinity to BD1 and BD2 of BRD4 with K d s of 90 and 28 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.