Learn More
Enzo Life Sciences Arv-825 (5mg). CAS: 1818885-28-7
SDP
Catalog No. 502013713
Encompass_Preferred
ARV-825 is a PROTAC targeting on BRD4, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand Pomalidomide, with Kds values of 90nM and 28nM for affinity to BD1 and BD2. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-3713 Supplier Enzo Life Sciences Supplier No. ENZCHM2580005
ARV-825 is a PROTAC targeting on BRD4, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand Pomalidomide, with Kds values of 90nM and 28nM for affinity to BD1 and BD2. Purity: ≥98%. Long Term Storage: -20°C.