Learn More
Aviva Systems Biology Corp ARV1 Antibody - middle region (ARP49756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARV1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARV1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Pig, Rabbit
Catalog No. 50-014-60170