Learn More
Aviva Systems Biology Corp ARX Antibody - middle region (ARP36824_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36824_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ARX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse ARX. Alternate names: ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56278