Learn More
Aviva Systems Biology Corp ARX Antibody - middle region (ARP31598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARX. Alternate names: ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRXS1, PRTS, CT121, EIEE1, MRX76, MRX87ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54276