Learn More
Selleck Chemical LLC CH-223191 10mg 301326-22-7
SDP
Supplier:  Selleck Chemical LLC S771110MG
Encompass_Preferred
CH-223191 is a potent and specific aryl hydrocarbon receptor (AhR) antagonist with IC50 of 30 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1332574