Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aryl Hydrocarbon Receptor Antibody (YA610) | Predicted band size: 96 kDa; Observed band size: 105 kDa | 10 UL
SDP

Catalog No. 5000396462
Encompass_Preferred

Aryl Hydrocarbon Receptor Antibody (YA610) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Aryl Hydrocarbon Receptor. It plays a critical role in cellular adaptation to environmental, dietary, microbial, and metabolic cues, influencing development, immunity, and cancer. For research use only.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Immunogen: Synthetic human Aryl Hydrocarbon Receptor peptide (AA range: 735-775)
  • Applications: WB (1:1000-1:2000), ICC/IF (1:100-1:500)
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage: -20°C for 1 year; avoid freeze/thaw cycles
  • Shipping: Blue ice

Catalog No. 50-003-96462 Supplier Medchemexpress LLC Supplier No. HYP8037810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aryl Hydrocarbon Receptor Antibody (YA610) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Aryl Hydrocarbon Receptor. It plays a critical role in cellular adaptation to environmental, dietary, microbial, and metabolic cues, influencing development, immunity, and cancer. For research use only.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Immunogen: Synthetic human Aryl Hydrocarbon Receptor peptide (AA range: 735-775)
  • Applications: WB (1:1000-1:2000), ICC/IF (1:100-1:500)
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, 0.05% Sodium Azide
  • Storage: -20°C for 1 year; avoid freeze/thaw cycles
  • Shipping: Blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.