Learn More
Medchemexpress LLC Aryl Hydrocarbon Receptor Antibody (YA610) | Predicted band size: 96 kDa; Observed band size: 105 kDa | 50 UL
SDP
Catalog No. 5000396248
Encompass_Preferred
Aryl Hydrocarbon Receptor Antibody (YA610) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting the Aryl Hydrocarbon Receptor. It is for research use only and not intended for patient use.
  • Rabbit-derived recombinant monoclonal antibody
  • Targets human Aryl Hydrocarbon Receptor
  • Validated for Western Blot and Immunocytochemistry/Immunofluorescence
  • Supplied as a liquid in TBS with BSA, glycerol, and sodium azide preservative
  • Store at -20°C for up to 1 year
  • Plays a role in development, immunity, and cancer
  • Regulates angiogenesis, hematopoiesis, and metabolism
  • Functions as a ligand-activated transcription factor
  • Subcellular localization in cytoplasm and nucleus
Catalog No. 50-003-96248 Supplier Medchemexpress LLC Supplier No. HYP8037850UL
Aryl Hydrocarbon Receptor Antibody (YA610) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting the Aryl Hydrocarbon Receptor. It is for research use only and not intended for patient use.
  • Rabbit-derived recombinant monoclonal antibody
  • Targets human Aryl Hydrocarbon Receptor
  • Validated for Western Blot and Immunocytochemistry/Immunofluorescence
  • Supplied as a liquid in TBS with BSA, glycerol, and sodium azide preservative
  • Store at -20°C for up to 1 year
  • Plays a role in development, immunity, and cancer
  • Regulates angiogenesis, hematopoiesis, and metabolism
  • Functions as a ligand-activated transcription factor
  • Subcellular localization in cytoplasm and nucleus