Learn More
Cayman Chemical Aryl Hydrocarbon Receptor Bioassay, 96 Well
SDP
Catalog No. 502945175
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for AhR activity. In addition to Reporter Cells, included are two optimized media for use during cell culture and in diluting the user's test samples, Luciferase Detection Reagent, and a cell culture-ready assay plate
Catalog No. 50-294-5175 Supplier Cayman Chemical Supplier No. 4103796 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for AhR activity. In addition to Reporter Cells, included are two optimized media for use during cell culture and in diluting the user's test samples, Luciferase Detection Reagent, and a cell culture-ready assay plate