Learn More
Cayman Chemical ArylpIperazIn 5k 10mg
SDP
Catalog No. 502916291
Encompass_Preferred
A FATP1 inhibitor (IC50 = 4.6 nM for the human protein); reduces neutral and polar lipid accumulation induced by linoleic acid in MDA-MB-231 cells at 12.5 µM; decreases ATP and BODIPY-palmitate levels in CD37-/- knockout BJAB cells but not in wild-type BJAB cells at 12.5 µM; induces cell death in MDA-MB-231, MCF-7, HCC1806, and BT474 cells at 12.5 µM
Catalog No. 50-291-6291 Supplier Cayman Chemical Supplier No. 3813510 MG
A FATP1 inhibitor (IC50 = 4.6 nM for the human protein); reduces neutral and polar lipid accumulation induced by linoleic acid in MDA-MB-231 cells at 12.5 µM; decreases ATP and BODIPY-palmitate levels in CD37-/- knockout BJAB cells but not in wild-type BJAB cells at 12.5 µM; induces cell death in MDA-MB-231, MCF-7, HCC1806, and BT474 cells at 12.5 µM