Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arylsulfatase A (ARSA) protein | >95.0% | 10 UG
SDP

Catalog No. 5000293037
Encompass_Preferred

Recombinant mouse arylsulfatase A (ARSA) supplied as a C-terminal His-tagged protein expressed in HEK293 cells. The enzyme hydrolyzes cerebroside sulfate and is provided at >95% purity in a 10 μg quantity for use in biochemical assays, enzymatic studies, antibody development, and as a positive control. Store and handle according to standard protein storage and reconstitution procedures.

  • Recombinant mouse protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied as a 10 μg quantity suitable for assay development.
  • Suitable for enzymatic assays, antibody generation, and assay controls.

Catalog No. 50-002-93037 Supplier Medchemexpress LLC Supplier No. HYP7549910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse arylsulfatase A (ARSA) supplied as a C-terminal His-tagged protein expressed in HEK293 cells. The enzyme hydrolyzes cerebroside sulfate and is provided at >95% purity in a 10 μg quantity for use in biochemical assays, enzymatic studies, antibody development, and as a positive control. Store and handle according to standard protein storage and reconstitution procedures.

  • Recombinant mouse protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied as a 10 μg quantity suitable for assay development.
  • Suitable for enzymatic assays, antibody generation, and assay controls.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.