Learn More
Medchemexpress LLC Arylsulfatase A (human recombinant) | 9068-68-2 | >95.0% | 50 UG
SDP
Catalog No. 5000292394
Encompass_Preferred
Recombinant human arylsulfatase A (ARSA) produced in HEK293 cells with a C-terminal His tag. The enzyme is lyophilized for stability, has an approximate molecular weight of 65 kDa, and is purified to >95% by reducing SDS-PAGE. It is intended for biochemical and cellular studies of sulfatide metabolism and enzymology.
  • Human recombinant enzyme expressed in HEK293 cells for human-relevant activity.
  • C-terminal His tag enables detection and affinity purification.
  • Lyophilized formulation improves shelf life and simplifies storage.
  • Purity greater than 95% by reducing SDS-PAGE for consistent performance.
  • Approximate molecular weight 65 kDa; suitable for SDS-PAGE and mass analysis.
  • Store lyophilized at -20°C; aliquot and freeze for extended storage.
  • Do not reconstitute to concentrations below 100 μg/mL in ddH2O.
  • Available in microgram pack sizes suitable for assay development.
Catalog No. 50-002-92394 Supplier Medchemexpress LLC Supplier No. HYP7608A50UG
Recombinant human arylsulfatase A (ARSA) produced in HEK293 cells with a C-terminal His tag. The enzyme is lyophilized for stability, has an approximate molecular weight of 65 kDa, and is purified to >95% by reducing SDS-PAGE. It is intended for biochemical and cellular studies of sulfatide metabolism and enzymology.
  • Human recombinant enzyme expressed in HEK293 cells for human-relevant activity.
  • C-terminal His tag enables detection and affinity purification.
  • Lyophilized formulation improves shelf life and simplifies storage.
  • Purity greater than 95% by reducing SDS-PAGE for consistent performance.
  • Approximate molecular weight 65 kDa; suitable for SDS-PAGE and mass analysis.
  • Store lyophilized at -20°C; aliquot and freeze for extended storage.
  • Do not reconstitute to concentrations below 100 μg/mL in ddH2O.
  • Available in microgram pack sizes suitable for assay development.