Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arylsulfonamide-based derivative (MPT0B390) | 1817802-18-8 | 98.8% | 375.40 | C17H17N3O5S | 5 MG
SDP

Catalog No. 5000221638
Encompass_Preferred

MPT0B390 is an arylsulfonamide-based small molecule HDAC inhibitor and TIMP3 inducer for research use. It shows antiproliferative activity and inhibition of tumor growth, metastasis, and angiogenesis. Identifiers include CAS 1817802-18-8 and formula C17H17N3O5S.

  • High purity (98.8%) suitable for research applications.
  • Molecular weight 375.40 g·mol-1.
  • Chemical formula C17H17N3O5S.
  • Available in solid and solution formats, including 5 mg solid packaging.
  • Provided with supporting documentation such as an SDS and analytical data.

Catalog No. 50-002-21638 Supplier Medchemexpress LLC Supplier No. HY1454265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MPT0B390 is an arylsulfonamide-based small molecule HDAC inhibitor and TIMP3 inducer for research use. It shows antiproliferative activity and inhibition of tumor growth, metastasis, and angiogenesis. Identifiers include CAS 1817802-18-8 and formula C17H17N3O5S.

  • High purity (98.8%) suitable for research applications.
  • Molecular weight 375.40 g·mol-1.
  • Chemical formula C17H17N3O5S.
  • Available in solid and solution formats, including 5 mg solid packaging.
  • Provided with supporting documentation such as an SDS and analytical data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.