Learn More
AdipoGen Arzanol (1 mg)
SDP
Supplier:  AdipoGen AGCN20500M001
Encompass_Preferred
Chemical. CAS: 32274-52-5. Formula: C22H26O7. MW: 402.4. Isolated from Helichrysum italicum. Anti-inflammatory compound. Potent dual inhibitor of pro-inflammatory transcription factors and inflammatory enzymes. Inhibitor of NFkappaB activation. Potent inhibitor of inducible microsomal prostaglandin E2 synthase-1 (mPGES-1), the terminal synthase responsible for the synthesis of the pro-tumorigenic prostaglandin E2 (PGE2). Antioxidant. 5-Lipoxygenase (5-LO) inhibitor. Antibacterial compound. HIV-1 replication inhibitor.
Catalog No. 50-205-3683