Learn More
TARGETMOL CHEMICALS INC AS-0141 5MG
SDP
Catalog No. 502581683
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AS-0141 (Cdc7-IN-6) is a potent Cdc7 kinase inhibitor (IC50=4 nM) extracted from patent WO2019165473A1(compound I- D) with anti-tumor activity. purity: 100%
Catalog No. 50-258-1683 Supplier TARGETMOL CHEMICALS INC Supplier No. T107285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AS-0141 (Cdc7-IN-6) is a potent Cdc7 kinase inhibitor (IC50=4 nM) extracted from patent WO2019165473A1(compound I- D) with anti-tumor activity. purity: 100%