Learn More
Abcam AS-041164, PI3K gamma inhibitor, 25MG
Supplier:  Abcam AB12089825MG
Encompass_Preferred
MW 249.24 g/mol, Purity >98%. Potent PI3K inhibitor with selectivity for PI3Kγ (IC₅₀ = 70 nM), compared to PI3Kα, β and δ (IC₅₀ values are 240 nM, 1.45 and 1.70 μM, respectively). Shows little or n o activity against 38 other common kinases. Blocks neutrophil migration. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7150