Learn More
TARGETMOL CHEMICALS INC AS-136A 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T621601MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. AS-136A is an orally active measles virus RNA-dependent RNA polymerase (RdRp) inhibitor with antiviral activity that inhibits measles virus and blocks viral RNA synthesis. Purity 99.96%
Catalog No. 50-287-9988