Learn More
Medchemexpress LLC AS 1411 (AGRO-100) | 301636-59-9 | 99.5% | 8272.31 | 1 MG
SDP
Catalog No. 5000359905
Encompass_Preferred
AS 1411 (AGRO-100) is an oligonucleotide aptamer that targets nucleoproteins. It inhibits tumor cell proliferation by affecting the activity of nucleoprotein-containing complexes. This aptamer can also serve as a carrier to precisely deliver nanoparticles, oligonucleotides, and small molecules to cancer cells.
  • Inhibits tumor cell proliferation
  • Reduces PRMT5 expression
  • Blocks binding of nucleoproteins to bcl-2 mRNA
  • Serves as a carrier for targeted delivery
  • Crosses the blood-brain barrier with low tissue toxicity
Catalog No. 50-003-59905 Supplier Medchemexpress LLC Supplier No. HY1470811MG
AS 1411 (AGRO-100) is an oligonucleotide aptamer that targets nucleoproteins. It inhibits tumor cell proliferation by affecting the activity of nucleoprotein-containing complexes. This aptamer can also serve as a carrier to precisely deliver nanoparticles, oligonucleotides, and small molecules to cancer cells.
  • Inhibits tumor cell proliferation
  • Reduces PRMT5 expression
  • Blocks binding of nucleoproteins to bcl-2 mRNA
  • Serves as a carrier for targeted delivery
  • Crosses the blood-brain barrier with low tissue toxicity