Learn More
Cayman Chemical AS-2444697 (hydrochloride), Purity: ≥95%, CAS: 1287665-60-4, 10 mg
SDP
Catalog No. 502989642
Encompass_Preferred
An IRAK4 inhibitor (IC50 = 21 nM); greater than 30-fold selective for IRAK4 over a panel of 146 kinases, but is less than 10-fold selective over PDGFRα, PDGFRβ, TrkA, TrkC, CLK1, Itk, and FLT3; inhibits IL-1β-induced IL-6 production and LPS-induced TNF-α production in cellular assays (IC50s = 250 and 47 nM, respectively); efficacious in rat models of adjuvant- and collagen-induced arthritis (ED50s = 2.7 and 1.6 mg/kg, respectively); reduces urinary protein excretion, the development of interstitial fibrosis and glomerulosclerosis, and renal mRNA expression of genes encoding IL-1β, IL-6, TNF-α, and CCL2 in the 5/6 nephrectomized rat model of chronic kidney disease from 0.3-3 mg/kg
Catalog No. 50-298-9642 Supplier Cayman Chemical Supplier No. 2950510 MG
An IRAK4 inhibitor (IC50 = 21 nM); greater than 30-fold selective for IRAK4 over a panel of 146 kinases, but is less than 10-fold selective over PDGFRα, PDGFRβ, TrkA, TrkC, CLK1, Itk, and FLT3; inhibits IL-1β-induced IL-6 production and LPS-induced TNF-α production in cellular assays (IC50s = 250 and 47 nM, respectively); efficacious in rat models of adjuvant- and collagen-induced arthritis (ED50s = 2.7 and 1.6 mg/kg, respectively); reduces urinary protein excretion, the development of interstitial fibrosis and glomerulosclerosis, and renal mRNA expression of genes encoding IL-1β, IL-6, TNF-α, and CCL2 in the 5/6 nephrectomized rat model of chronic kidney disease from 0.3-3 mg/kg