Learn More
TARGETMOL CHEMICALS INC AS-252424 10MG
SDP
Catalog No. 502576573
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS-252424 is a potent and selective inhibitor of PI3K(gamma) with IC50 of 33 nM; >10 fold selectivity for PI3K(gamma) versus PI3K(alpha). purity: 99%
Catalog No. 50-257-6573 Supplier TARGETMOL CHEMICALS INC Supplier No. T620810MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AS-252424 is a potent and selective inhibitor of PI3K(gamma) with IC50 of 33 nM; >10 fold selectivity for PI3K(gamma) versus PI3K(alpha). purity: 99%