Learn More
Enzo Life Sciences AS-252424 (1mg). CAS: 900515-16-4
SDP
Catalog No. 502008587
Encompass_Preferred
Potent and selective phosphoinositide 3-kinase (PI(3)K) p110γ inhibitor shown to be selective for class IB PI(3)K-mediated cellular effects. Alternative name: 5-[5-(4-Fluoro-2-hydroxyphenyl)-furan-2-ylmethylene)]-thiazolidine-2,4-dione. Purity: ≥98% (1H-NMR). Solubility: Soluble in 100% ethanol (10mg/ml), DMSO (20mg/ml) or dimethyl formamide; also soluble in a 1:1 solution of DMSO:PBS, pH 7.4, (0.5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8587 Supplier Enzo Life Sciences Supplier No. ALX270465M001
Potent and selective phosphoinositide 3-kinase (PI(3)K) p110γ inhibitor shown to be selective for class IB PI(3)K-mediated cellular effects. Alternative name: 5-[5-(4-Fluoro-2-hydroxyphenyl)-furan-2-ylmethylene)]-thiazolidine-2,4-dione. Purity: ≥98% (1H-NMR). Solubility: Soluble in 100% ethanol (10mg/ml), DMSO (20mg/ml) or dimethyl formamide; also soluble in a 1:1 solution of DMSO:PBS, pH 7.4, (0.5mg/ml). Long Term Storage: -20°C.