Learn More
Apexbio Technology LLC AS-252424, 5mg. Cas: 900515-16-4 MFCD: MFCD12024675. PI3Kγ inhibitor,novel and potent
SDP
Supplier:  Apexbio Technology LLC B21805
Encompass_Preferred
AS-252424 is a potent, selective and novel small-molecule PI3K inhibitor (IC50= 30 nM) PI3K (phosphatidylinositol-4,5-bisphosphate 3-kinase) is a family of enzymes involved in cellular functions such as cell growth, proliferation, differentiation, motility, survival and intracellular trafficking. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5326