Learn More
Medchemexpress LLC HY-13531 5mg Medchemexpress, AS-604850 CAS:648449-76-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135315MG
Encompass_Preferred
Medchemexpress, HY-13531 5mg AS-604850 CAS:648449-76-7 AS-604850 is a potent, selective and ATP-competitive PI3Kγ inhibitor with an IC50 value of 0.25 μM and a Ki value of 0.18 μM. AS-604850 shows isoform selective inhibitor of PI3Kγ with over 30-fold selectivity for PI3Kδ and β, and 18-fold selectivity over PI3Kα, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9873