Learn More
Enzo Life Sciences AS-604850 (5mg). CAS: 648449-76-7
SDP
Supplier:  Enzo Life Sciences ALX270461M005
Encompass_Preferred
Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ). Exhibits selectivity over other PI(3)K isoforms. Alternative name: 5-(2,2-Difluoro-benzo[1,3]dioxol-5-ylmethylene)-thiazolidine-2,4-dione. Purity: ≥98% (HPLC (UV)). Appearance: Off-white to yellow solid. Solubility: Soluble in acetonitrile (6mg/ml), DMSO (50mg/ml), methanol (9mg/ml) or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8584