Learn More
Enzo Life Sciences AS-605240 (5mg). CAS: 648450-29-7
SDP
Catalog No. 502008585
Encompass_Preferred
Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ). Exhibits selectivity over other PI(3)K isoforms. Alternative name: 5-Quinoxalin-6-ylmethylene-thiazolidine-2,4-dione. Purity: ≥98% (HPLC). Appearance: Pink to reddish brown solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8585 Supplier Enzo Life Sciences Supplier No. ALX270462M005
Add to Cart
Add to Cart
Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ). Exhibits selectivity over other PI(3)K isoforms. Alternative name: 5-Quinoxalin-6-ylmethylene-thiazolidine-2,4-dione. Purity: ≥98% (HPLC). Appearance: Pink to reddish brown solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C.