Learn More
Selleck Chemical LLC AS-605240 5mg 648450-29-7
SDP
Catalog No. 508477
Encompass_Preferred
AS-605240 selectively inhibits PI3K? with IC50 of 8 nM, over 30-fold and 7.5-fold more selective for PI3K? than PI3K?/? and PI3K? in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-847-7 Supplier Selleck Chemical LLC Supplier No. S14105MG
AS-605240 selectively inhibits PI3K? with IC50 of 8 nM, over 30-fold and 7.5-fold more selective for PI3K? than PI3K?/? and PI3K? in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.