Learn More
Medchemexpress LLC AS-605240 potassium salt | 648450-29-7 | 99.4% | 257.27 g/mol | C12H7N3O2S | 1 ML
SDP
Catalog No. 5000213317
Encompass_Preferred
AS-605240 is a selective, orally active inhibitor of PI3Kγ used in research to probe PI3Kγ-dependent signaling and inflammatory responses. It inhibits PI3Kγ with an IC50 of 8 nM and a Ki of 7.8 nM and is supplied as a 10 mM solution in DMSO (1 mL) for use in cellular and in vivo studies.
  • Selective PI3Kγ inhibition (IC50 8 nM).
  • High binding affinity (Ki 7.8 nM).
  • Suitable for cellular and in vivo pharmacology studies.
  • Delivered as 10 mM in DMSO, 1 mL format.
  • High purity, suitable for research applications.
Catalog No. 50-002-13317 Supplier Medchemexpress LLC Supplier No. HY1010910MM 1ML
AS-605240 is a selective, orally active inhibitor of PI3Kγ used in research to probe PI3Kγ-dependent signaling and inflammatory responses. It inhibits PI3Kγ with an IC50 of 8 nM and a Ki of 7.8 nM and is supplied as a 10 mM solution in DMSO (1 mL) for use in cellular and in vivo studies.
  • Selective PI3Kγ inhibition (IC50 8 nM).
  • High binding affinity (Ki 7.8 nM).
  • Suitable for cellular and in vivo pharmacology studies.
  • Delivered as 10 mM in DMSO, 1 mL format.
  • High purity, suitable for research applications.