Learn More
Cayman Chemical AS-605240potasIum salt 10mg
SDP
Catalog No. 502892569
Encompass_Preferred
An orally active inhibitor of PI3-kinase γ that suppresses joint inflammation in mouse models of rheumatoid arthritis; inhibits human recombinant PI3Kγ, α, β, and δ in an ATP-competitive manner with IC50 values of 8, 60, 270, and 300 nM, respectively
Catalog No. 50-289-2569 Supplier Cayman Chemical Supplier No. 900098010 MG
An orally active inhibitor of PI3-kinase γ that suppresses joint inflammation in mouse models of rheumatoid arthritis; inhibits human recombinant PI3Kγ, α, β, and δ in an ATP-competitive manner with IC50 values of 8, 60, 270, and 300 nM, respectively