Learn More
Selleck Chemical LLC AS-8351-5mg
SDP
Supplier:  Selleck Chemical LLC S57725MG
Encompass_Preferred
AS-8351 is a histone demethylase inhibitor that could induces reprogramming of human fetal lung fibroblasts into functional cardiomyocytes.
Catalog No. 50-239-0015