Learn More
Medchemexpress LLC AS-99 TFA | 00-00-0 | 99.2% | C29H31F6N5O5S2 | 10MG
SDP
Catalog No. 5000261557
Encompass_Preferred
AS-99 TFA is the trifluoroacetic acid salt of AS-99, a potent and selective inhibitor of the ASH1L histone methyltransferase. It exhibits anti-leukemic activity by blocking proliferation, inducing apoptosis and differentiation, and downregulating MLL fusion target genes; intended for mechanistic epigenetics and preclinical leukemia research.
  • Potent and selective ASH1L histone methyltransferase inhibition (IC50 ≈ 0.79 μM).
  • Demonstrated anti-leukemic activity in cellular and in vivo models.
  • High purity (99.2%).
  • Available as a TFA salt and supplied as solid or DMSO solutions.
  • Includes manufacturer documentation: data sheet, COA, and SDS for quality and safety.
Catalog No. 50-002-61557 Supplier Medchemexpress LLC Supplier No. HY141429A10MG
Add to Cart
Add to Cart
AS-99 TFA is the trifluoroacetic acid salt of AS-99, a potent and selective inhibitor of the ASH1L histone methyltransferase. It exhibits anti-leukemic activity by blocking proliferation, inducing apoptosis and differentiation, and downregulating MLL fusion target genes; intended for mechanistic epigenetics and preclinical leukemia research.
  • Potent and selective ASH1L histone methyltransferase inhibition (IC50 ≈ 0.79 μM).
  • Demonstrated anti-leukemic activity in cellular and in vivo models.
  • High purity (99.2%).
  • Available as a TFA salt and supplied as solid or DMSO solutions.
  • Includes manufacturer documentation: data sheet, COA, and SDS for quality and safety.