Learn More
Medchemexpress LLC AS-99 TFA | 3115178-55-4 | 99.2% | C29H31F6N5O5S2 | 100 MG
SDP
Catalog No. 5000354792
Encompass_Preferred
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor with anti-leukemic activity. It blocks cell proliferation, induces apoptosis and differentiation, downregulates MLL fusion target genes, and reduces the leukemia burden in vivo.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Exhibits anti-leukemic activity
  • Blocks cell proliferation
  • Induces apoptosis and differentiation
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Shows over 100-fold selectivity towards ASH1L
Catalog No. 50-003-54792 Supplier Medchemexpress LLC Supplier No. HY141429A100MG
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor with anti-leukemic activity. It blocks cell proliferation, induces apoptosis and differentiation, downregulates MLL fusion target genes, and reduces the leukemia burden in vivo.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Exhibits anti-leukemic activity
  • Blocks cell proliferation
  • Induces apoptosis and differentiation
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Shows over 100-fold selectivity towards ASH1L