Learn More
Medchemexpress LLC AS-99 TFA | 99.1% | C29H31F6N5O5S2 | 1 MG
SDP
Catalog No. 5000353431
Encompass_Preferred
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor (IC50 of 0.79 μM, Kd of 0.89 μM) with anti-leukemic activity. It functions by blocking cell proliferation, inducing apoptosis and differentiation, downregulating MLL fusion target genes, and reducing the leukemia burden in vivo.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Demonstrates anti-leukemic activity
  • Blocks cell proliferation
  • Induces apoptosis and differentiation
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Exhibits over 100-fold selectivity towards ASH1L over 20 other histone methyltransferases
  • Suppresses MLL fusion driven transcriptional programs
  • Leads to dose-dependent downregulation of canonical MLL fusion target genes
Catalog No. 50-003-53431 Supplier Medchemexpress LLC Supplier No. HY141429A1MG
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor (IC50 of 0.79 μM, Kd of 0.89 μM) with anti-leukemic activity. It functions by blocking cell proliferation, inducing apoptosis and differentiation, downregulating MLL fusion target genes, and reducing the leukemia burden in vivo.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Demonstrates anti-leukemic activity
  • Blocks cell proliferation
  • Induces apoptosis and differentiation
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Exhibits over 100-fold selectivity towards ASH1L over 20 other histone methyltransferases
  • Suppresses MLL fusion driven transcriptional programs
  • Leads to dose-dependent downregulation of canonical MLL fusion target genes