Learn More
Medchemexpress LLC AS-99 TFA | 99.2% | C29H31F6N5O5S2 | 50 MG
SDP
Catalog No. 5000355145
Encompass_Preferred
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor with anti-leukemic activity. It blocks cell proliferation, induces apoptosis and differentiation, downregulates MLL fusion target genes, and reduces leukemia burden in vivo. The compound exhibits over 100-fold selectivity towards ASH1L, showing no significant inhibition on a panel of 20 other histone methyltransferases at 50 μM.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Blocks cell proliferation and induces apoptosis
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Exhibits over 100-fold selectivity towards ASH1L
  • Favorable exposure in plasma with a half-life of approximately 5-6 hours
Catalog No. 50-003-55145 Supplier Medchemexpress LLC Supplier No. HY141429A50MG
AS-99 TFA is a first-in-class, potent, and selective ASH1L histone methyltransferase inhibitor with anti-leukemic activity. It blocks cell proliferation, induces apoptosis and differentiation, downregulates MLL fusion target genes, and reduces leukemia burden in vivo. The compound exhibits over 100-fold selectivity towards ASH1L, showing no significant inhibition on a panel of 20 other histone methyltransferases at 50 μM.
  • Potent and selective ASH1L histone methyltransferase inhibitor
  • Blocks cell proliferation and induces apoptosis
  • Downregulates MLL fusion target genes
  • Reduces leukemia burden in vivo
  • Exhibits over 100-fold selectivity towards ASH1L
  • Favorable exposure in plasma with a half-life of approximately 5-6 hours