Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC As-99 trifluoroacetate | 99.2% | 707.71 | C29H31F6N5O5S2 | 5 MG
SDP

Catalog No. 5000222196
Encompass_Preferred

AS-99 trifluoroacetate is the trifluoroacetic acid (TFA) salt of AS-99, a potent and selective inhibitor of the ASH1L (KMT2H) histone methyltransferase. It is supplied as a research reagent for biochemical and cellular studies of epigenetic regulation and anti-leukemic activity.

  • TFA salt form to aid solubility and handling.
  • High purity (99.15%).
  • Molecular formula C29H31F6N5O5S2 and molecular weight 707.71 Da.
  • Available as 1-100 mg solid quantities and 10 mM in 1 mL DMSO solutions.
  • Intended for biochemical and cellular assay use.

Catalog No. 50-002-22196 Supplier Medchemexpress LLC Supplier No. HY141429A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AS-99 trifluoroacetate is the trifluoroacetic acid (TFA) salt of AS-99, a potent and selective inhibitor of the ASH1L (KMT2H) histone methyltransferase. It is supplied as a research reagent for biochemical and cellular studies of epigenetic regulation and anti-leukemic activity.

  • TFA salt form to aid solubility and handling.
  • High purity (99.15%).
  • Molecular formula C29H31F6N5O5S2 and molecular weight 707.71 Da.
  • Available as 1-100 mg solid quantities and 10 mM in 1 mL DMSO solutions.
  • Intended for biochemical and cellular assay use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.