Learn More
Bachem Chlorotoxin Acetate
SDP
Supplier:  Bachem 4079411.1000
Encompass_Preferred
Chlorotoxin Acetate, H-Met-Cys-Met-Pro-Cys-Phe-Thr-Thr-Asp-His-Gln-Met-Ala-Arg-Lys-Cys-Asp-Asp-Cys-Cys-Gly-Gly-Lys-Gly-Arg-Gly-Lys-Cys-Tyr-Gly-Pro-Gln-Cys-Leu-Cys-Arg-NH2, 163515-35-3, C158H249N53O47S11, Mr 3995.77, 1mg. Cltx (Egyptian scorpion). As chlorotoxin has shown a remarkable affinity for binding to tumor cells, it has been conjugated to cancer drugs or radionuclides for diagnostic or therapeutic uses.
Catalog No. 50-194-6733